• USA Home
  • R8631 - Bcl I from Bacillus caldolyticus

EMAIL THIS PAGE TO A FRIEND
R8631 Sigma

Bcl I from Bacillus caldolyticus

Restriction Enzyme

Purchase

Properties

Related Categories 3.1.x.x Acting on esters, 3.x.x.x Hydrolases, Application Index, Biochemicals and Reagents, Enzyme Class Index,
grade   for molecular biology
form   buffered aqueous glycerol solution
concentration   10,000 units/mL
shipped in   wet ice
storage temp.   −20°C

Description

Other Notes

Supplied with 10x Restriction Enzyme Buffer SM (B3158).

Cleavage activity: Bcl I will only partially cleave DNA isolated from E. coli strains that have the dam methylase (dam+ strains).
Comment: Bcl I is optimally active at 50°C.

Specificity

Recognition sequence: 5′-T/GATCA-3′
Cutting results: a 2-10-fold Bcl I overdigestion of 1 μg λ DNA substrate results in 100% cutting.
Heat inactivation: This enzyme cannot be heat inactivated at 65 °C for 15 minutes.

Physical form

Solution in 20 mM Tris-HCl, pH 8.0, 0.1 mM EDTA, 200 mM NaCl, 10 mM 2-Mercaptoethanol, 0.2 % Triton X-100 (v/v), 50% glycerol (v/v) at 4°C

Application

BclI is a restriction endonuclease used in molecular biology applications to cut DNA at the recognition sequence 5′-T/GATCA-3′, resulting in fragments with 5′-cohesive termini.

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Articles

Restriction Enzyme Buffer Reference

Sigma® Life Science’s restriction enzyme collection has been optimized for digestion using five unique buffers. Reference below the enzyme activity in each buffer (table 1), and the preferred buffer ...
Keywords: Buffers, Digestions, Enzyme activity, PAGE, Solid phase extractions

Protocols

Restriction Enzyme Digest Protocol

Sigma® Life Science’s restriction enzyme collection has been optimized for digestion using five unique buffers. When digesting DNA using a single enzyme, use the buffer supplied with the enzyme (also...
Keywords: Buffers, Digestions, Purification

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

incubation buffer

Product #

Description

Add to Cart

B3158 Restriction Enzyme Buffer SM, 10 ×

related product

Product #

Description

Add to Cart

LIG2 QuickLink DNA Ligation Kit, joins blunt-end and sticky-end DNA fragments
R0260 BamH I from Bacillus amyloliquefaciens H, Restriction Enzyme
R7260 Xba I from Xanthomonas badrii, Restriction Enzyme
D2886 DNA Ligase from T4-infected Escherichia coli, buffered aqueous glycerol solution
R1137 Hind III from Haemophilus influenzae, Restriction Enzyme
R2756 EcoR V from Escherichia coli, buffered aqueous glycerol solution

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?