• USA Home
  • R8636 - Monoclonal Anti-Rat Kappa & Lambda Light Chains antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
R8636 Sigma

Monoclonal Anti-Rat Kappa & Lambda Light Chains antibody produced in mouse

clone RT-39/RL-6, ascites fluid

Synonym: Monoclonal Anti-Rat Kappa & Lambda Light Chains

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Primary Antibodies, RAE-RAX, Rat Secondary Antibodies and Conjugates,
biological source   mouse
antibody form   ascites fluid
clone   RT-39/RL-6, monoclonal
mol wt   antigen mol wt 25 kDa
contains   15 mM sodium azide
application(s)   immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
  immunohistochemistry (frozen sections): suitable
  indirect ELISA: 1:1,000
isotype   IgG1/IgG2a
conjugate   unconjugated
shipped in   dry ice
storage temp.   −20°C

Description

General description

The product is a mixture derived from hybridomas RT-39 and RL-6, secreting monoclonal antibodies against rat κ and λ light chains, respectively.

The product recognizes epitopes located on the rat κ and λ light chains on the various rat immunoglobulin classes and subclasses. The epitope recognized on the κ light chains is shared by both the 1a and 1b allotypes of this molecule. The antibody detects the κ and λ light chains derived from normal serum immunoglobulins or from myeloma proteins. Weak cross-reaction is observed with guinea pig immunoglobulins but not with human, bovine, cat, chicken, dog, goat, horse, mouse, pig, rabbit or sheep immunoglobulins.

Price and Availability


Antibody Explorer

Explore with Confidence

Amplified Detection
Safety & Documentation
Protocols & Articles

Articles

How to Choose a Secondary Antibody

The following information is provided to help you decide which secondary antibody may be best for your particular application.
Keywords: Amplification, Enzyme-linked immunosorbent assay, Flow cytometry, Immobilization, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western blot

Secondary Antibodies, Conjugates and Kits

Secondary antibodies are polyclonal or monoclonal antibodies that bind to primary antibodies or antibody fragments, such as the Fc or Fab regions. They are typically labeled with probes that make the...
Keywords: Absorption, Adsorption, Amplification, Bacterial conjugations, Cancer, Digestions, Electrophoresis, Enzyme-linked immunosorbent assay, Flow cytometry, Gel electrophoresis, Gene expression, Hormones, Immobilization, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunology, Immunoprecipitation, Infrared spectroscopy, Microbiology, Microscopy, Purification, Vitamins, Western blot

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?