• USA Home
  • R8886 - Monoclonal Anti-CD4−Quantum Red antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
R8886 Sigma

Monoclonal Anti-CD4−Quantum Red antibody produced in mouse

clone Q4120, purified immunoglobulin, buffered aqueous solution

Synonym: Monoclonal Anti-CD4

Properties

Related Categories Alphabetical Index, Analytical Cytology, Antibodies, Antibodies for Cell Biology, Antibodies for Immune Cell Signaling and Blood,
species reactivity   human
application(s)   flow cytometry: 10 μL using 1 × 106 cells
clone   Q4120, monoclonal
antibody form   purified immunoglobulin
form   buffered aqueous solution
isotype   IgG1
mol wt   antigen mol wt 59 kDa
shipped in   wet ice
storage temp.   2-8°C
Gene Information   human ... CD4(920)
biological source   mouse
conjugate   Quantum Red® conjugate

Description

Immunogen

CD4-transfected mouse T-cell hybridoma, 3DT, followed by CD4+ human T-cell CEM cells.

Antigen Background

CD4 is a single chain transmembraneous glycoprotein from the immunoglobulin superfamily. It is expressed on the helper/inducer T subset, on most medullary thymocytes, on microglial, dendritic and on some malignancies of T cell origin. The antigen binds to MHC class II molecules and is associated with p56lck protein tyrosine kinase.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide

Specificity

Recognizes the CD4. The epitope recognized by the Q4120 clone is located on 1-183 aa and is sensitive to formalin fixation and paraffin embedding. 5th Workshop: code no. CD04.11

Preparation Note

Quantum Red is a tandem fluorescent dye created by coupling R-phycoerythrin with Cy5 reactive dye. This red dye is efficiently excited at 488 nm and emits at 670 nm.

Legal Information

Quantum Red is a trademark of Sigma-Aldrich Co. LLC

Texas Red is a registered trademark of Life Technologies

Price and Availability


Antibody Explorer

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?