• USA Home
  • R9375 - D-Ribulose o-nitrophenylhydrazone

EMAIL THIS PAGE TO A FRIEND
R9375 Sigma

D-Ribulose o-nitrophenylhydrazone

  • CAS Number 6155-41-5

  • Empirical Formula (Hill Notation) C11H15N3O6

  • Molecular Weight 285.25

  •  MDL number MFCD00135011

  •  PubChem Substance ID 24899413

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates P-Z, Monosaccharide More...
InChI Key   BXKCHSGKQBDYBI-MDWZMJQESA-N

Description

Packaging

Instructions for isolating D-ribulose for use as a chromatographic standard accompany each package.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?