• USA Home
  • R9508 - Vial Rack, Anodized Aluminum
EMAIL THIS PAGE TO A FRIEND
R9508 Sigma
Vial Rack, Anodized Aluminum
Purchase
Properties
Related Categories Cryo Containers, Cryoware, Labware More...
Description
General description
24-position rack designed specifically for use with freezing vials. External ribs on vials lock into serrated holes in rack, allowing single-handed opening and closing of screw caps.
Rack is autoclavable and stackable, and has an adjustable shelf to accommodate different size vials.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
CLS430525 Corning® cryogenic boxes, racks & trays, Rack and tray holds 30 vials
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?