• USA Home
  • R9890 - Floating microcentrifuge tube rack
EMAIL THIS PAGE TO A FRIEND
R9890 Sigma
Floating microcentrifuge tube rack
black
Purchase
Properties
Related Categories Floating microcentrifuge tube rack, Labware, Manufacturer Browser, Microcentrifuge Tube Racks, Nalgene,
material   polypropylene
description   Autoclavable
  Holds 0.5 mL tubes in a 4 × 4 array
mfr. no.   Nalgene
size   103 mm × 103 mm × 65 mm
color   black
Description
General description
On benchtop, legs prevent tubes from touching surface. In water bath, racks will float with a full load of tubes.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?