• USA Home
  • RAB0883 - Rat Ngf / Beta-nerve Growth Factor ELISA Kit

EMAIL THIS PAGE TO A FRIEND
RAB0883 Sigma

Rat Ngf / Beta-nerve Growth Factor ELISA Kit

for cell and tissue lysates

Purchase

Properties

Related Categories Antibodies, Antibody Kits, ELISA Kits, Rat, Sandwich ELISAs More...
species reactivity   rat
application(s)   ELISA: suitable
assay range   inter-assay cv: <10 %
intra-assay cv: <12 %
shipped in   wet ice
storage temp.   −20°C
Gene Information   rat ... Ngf(310738)

Description

Application

Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

General description

The antibody pair provided in this kit detects Rat beta-NGF in serum, plasma, cell culture supernatants.

Price and Availability


Antibody Explorer

Explore with Confidence

Making Headway

Kit component only

Description

Product #

Add to Cart

Rat Ngf Antibody-coated ELISA Plate (Item A) SDS    
Rat Ngf Protein Standard, Lyophilized (Item C) SDS    
Biotinylated Rat Ngf Detection Antibody (Item F) SDS    
HRP-Streptavidin (Item G) SDS    
ELISA Colorimetric TMB Reagent (HRP Substrate, Item H) SDS    
ELISA Stop Solution (Item I) SDS    
ELISA 5X Assay/Sample Diluent Buffer E (Item E2) SDS    
ELISA 1X Assay/Sample Diluent Buffer A (Item D1) SDS    
20X Wash Buffer (Item B) SDS    
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Warning
Hazard statements 
RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles

Protocols

ELISA Protocols

NOTE: ALL incubations and wash steps must be performed under gentle rocking or rotation (~1‐2 cycles/sec).
Keywords: Cell culture, Phosphorylations, Size-exclusion chromatography

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?