• USA Home
  • RAB0914 - Rat Tnfrsf1b / Tumor Necrosis Factor Receptor Superfamily Member 1B ELISA Kit

EMAIL THIS PAGE TO A FRIEND
RAB0914 Sigma

Rat Tnfrsf1b / Tumor Necrosis Factor Receptor Superfamily Member 1B ELISA Kit

Purchase

Properties

Related Categories Antibodies, Antibody Kits, ELISA Kits, Rat, Sandwich ELISAs More...
species reactivity   rat
application(s)   ELISA: suitable
assay range   inter-assay cv: <10 %
intra-assay cv: <12 %
shipped in   wet ice
storage temp.   −20°C
Gene Information   rat ... Tnfrsf1b(156767)

Description

Application

Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

General description

The antibody pair provided in this kit recognizes Rat Tumor Necrosis Factor Receptor Superfamily Member 1B.

Price and Availability


Antibody Explorer

Explore with Confidence

Direct Detect Infrared Spectrometer

Kit component only

Description

Product #

Add to Cart

Rat Tnfrsf1b Antibody-coated ELISA Plate (Item A) SDS    
Rat Tnfrsf1b Protein Standard, Lyophilized (Item C) SDS    
Biotinylated Rat Tnfrsf1b Detection Antibody (Item F) SDS    
HRP-Streptavidin (Item G) SDS    
ELISA Colorimetric TMB Reagent (HRP Substrate, Item H) SDS    
ELISA Stop Solution (Item I) SDS    
20X Wash Buffer (Item B) SDS    
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Warning
Hazard statements 
RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles

Protocols

ELISA Protocols

NOTE: ALL incubations and wash steps must be performed under gentle rocking or rotation (~1‐2 cycles/sec).
Keywords: Cell culture, Phosphorylations, Size-exclusion chromatography

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?