• USA Home
  • RDCL3 - Director-Ready pFLAG-CMV-2 Set

EMAIL THIS PAGE TO A FRIEND
RDCL3 Sigma

Director-Ready pFLAG-CMV-2 Set

Purchase

Properties

shipped in   wet ice
storage temp.   −20°C

Description

Components

pFLAG-CMV2
Verification primer-MF
Verification primer-MR

General description

Contains dephoshporylated- Hind III/Bgl II digested Director-Ready pFLAG-CMV2 for transient, cytoplasmic expression of N-terminal FLAG® fusion proteins in mammalian cells.

Legal Information

Director-Ready is a trademark of Sigma-Aldrich Co. LLC

FLAG is a registered trademark of Sigma-Aldrich Co. LLC

pFLAG-CMV is a trademark of Sigma-Aldrich Co. LLC

pFLAG-CMV2 is a trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?