• USA Home
  • RDCLIG1 - Director-Ready pFLAG-MAC Set

EMAIL THIS PAGE TO A FRIEND
RDCLIG1 Sigma

Director-Ready pFLAG-MAC Set

Purchase

Properties

form   buffered aqueous solution
shipped in   wet ice
storage temp.   −20°C

Description

Components

pFLAG-MAC
Verification primer-BF
Verification primer-MF

General description

Contains dephosphorylated-Hind III/Bgl II digested Director-Ready pFLAG-MAC for cytoplasmic expression of N-terminal FLAG® fusion proteins in E. coli.

Legal Information

Director-Ready is a trademark of Sigma-Aldrich Co. LLC

FLAG is a registered trademark of Sigma-Aldrich Co. LLC

pFLAG-MAC is a trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?