• USA Home
  • RPROBE1 - RadProbe Second-Strand Probe Synthesis Kit
EMAIL THIS PAGE TO A FRIEND
RPROBE1 Sigma
RadProbe Second-Strand Probe Synthesis Kit
Purchase
Properties
storage temp.   −20°C
Description
General description
RadProbe� Second Strand Synthesis Kit has been designed to generate second strand DNA probes with enhanced specific activity. Using DNA Polymerases rather than Reverse Transcriptases allows for the incorporate labeled nucleotides yields probes with highly enhanced specific activities1. Probes are produced by incorporating radiolabeled nucleotides (not included) in a random primed reaction from cDNA templates. The kit contains reagents, enzymes and protocols for first strand DNA synthesis and second strand labeling from polyA RNA (total or mRNA). RadProbe includes all the necessary reagents sufficient for 25 labeling reactions.
Legal Information
RadProbe is a trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Peer-Reviewed Papers
15
References
Set your institution to view full text papers.
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?