• USA Home
  • S0504 - Staphylococcal dried cells from Staphylococcus aureus

EMAIL THIS PAGE TO A FRIEND
S0504 Sigma

Staphylococcal dried cells from Staphylococcus aureus

Purchase

Properties

Related Categories Biochemicals and Reagents, Cells, Microbiology, Microorganisms, Protein Extracts,
sterility   Not processed or packaged aseptically
storage temp.   −20°C

Description

Application

(Not intended for use as a starter culture)

Contains clumping factor for use in detecting fibrin/fibrinogen degradation products in the Staphylococcal Clumping Test.

Preparation Note

Prepared from Strain Newman D2C.
Produced in pure culture.

Quality

Without additives

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 3462 6.1 / PGI
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?