• USA Home
  • S0889 - Sodium dithiophosphate Tribasic: hydrate

EMAIL THIS PAGE TO A FRIEND
S0889 Sigma

Sodium dithiophosphate Tribasic: hydrate

~95%

Synonym: Trisodium dithiophosphate

Purchase

Properties

assay   ~95%
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?