S0895 Sigma

Screens for CD-1

size 50 mesh

DOWNLOAD MSDS (PDF)

Synonym: cell sieve, mesh screen, screen mesh

Properties

Related Categories Cell Culture Supplies, Cell Dissociation Supplies, Labware More...
size   50 mesh

Description

Legal Information

CD-1 is a trademark of Sigma-Aldrich Co. LLC

Price and Availability

Customers Also Viewed

Cell dissociation sieve - tissue grinder kit

tissue grinder homogenizer kit, stainless steel, autoclavable, 1/kit

tightens screens in place for the CD-1 tissue grinder

size 85 mL

Screens for CD-1<SUP>™</SUP>

size 40 mesh

Screens for CD-1<SUP>™</SUP>

size 60 mesh

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?