• USA Home
  • S1042 - Starburst® dendrimer Generation 2

EMAIL THIS PAGE TO A FRIEND
S1042 Sigma

Starburst® dendrimer Generation 2

solution

Purchase

Properties

form   solution

Description

Legal Information

Starburst is a registered trademark of Dendritic NanoTechnologies, Inc.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 1230 3/PG 2
Flash Point(F) 
52 °F
Flash Point(C) 
11 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?