• USA Home
  • S1062 - 2-O-Sulfatase from Flavobacterium heparinum

EMAIL THIS PAGE TO A FRIEND
S1062 Sigma

2-O-Sulfatase from Flavobacterium heparinum

10-40 (units/ml)

Purchase

Description

General description

Hydrolyses the terminal 2-O-sulphate on the unsaturated di- and oligo-saccharides produced by the action of lyases on sulphated glycosaminoglycans.

Unit Definition

One unit will hydrolyze heparin disaccharide I-P (H9401) to yield 1.0 micromole of disaccharide II-P (δ-UA-GlcNCOEt-6S) per minute at pH 7.3 at 25 °C.

Physical form

Solution contains 10 mM tris-acetate, pH 7.3, 80 mM NaCl, 10 mM potassium phosphate, 0.2% BSA.

Application

2-O-Sulfatase from Flavobacterium heparinum has been used in a study to determine that oversulfated chondroitin sulfate is a contaminant in heparin associated with adverse clinical events. 2-O-Sulfatase from Flavobacterium heparinum has also been used in a study to determine the structure of oligosaccharides prepared from acharan sulfate.

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?