• USA Home
  • S10SM - Spectrophotometer cuvettes, silica (quartz)
EMAIL THIS PAGE TO A FRIEND
S10SM Sigma
Spectrophotometer cuvettes, silica (quartz)
Semi-micro with PTFE cover, volume ≥1 mL
Synonym: cuvette quartz, cuvette, cuvettes quartz
Purchase
Properties
Related Categories Labware, Spectroscopy Supplies, Spectroscopy, UV More...
material   quartz
label   A
volume   ≥1 mL
Description
General description
All spectrophotometer cuvettes are sold singly, not in pairs. With modern precision manufacturing methods, it is no longer necessary to pre-match cuvettes.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?