• USA Home
  • S1276 - Sodium borohydride tablets400 mg naBh4 per tablet

EMAIL THIS PAGE TO A FRIEND
S1276 Sigma

Sodium borohydride tablets400 mg naBh4 per tablet

Purchase

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 1426 4.3 / PGI
WGK Germany 
2
RTECS 
ED3325000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?