• USA Home
  • S1312 - Spectrophotometry and Spectrofluorimetry: A Practical Approach, 2nd ed.
EMAIL THIS PAGE TO A FRIEND
S1312 Sigma
Spectrophotometry and Spectrofluorimetry: A Practical Approach, 2nd ed.
  •  ISBN-10 0-19-963812-8
  •  ISBN-13 978-0-19-963812-3
Purchase
Properties
Related Categories Books and Software, Labware, Spectroscopy Books More...
publication info   M. Gore, ed., Oxford University Press, 2000, 384 pp., soft cover
mfr. no.   Oxford University Press
Description
General description
Optical spectroscopy underpins the day to day operations of most laboratories in the chemical, biological and medical sciences. This edition contains substantially updated and new chapters addressing the principles of the more common applications. In all chapters the emphasis is placed upon the practical aspects, with protocols for test experiments. Other chapters are included to introduce subjects that have traditionally depended upon spectroscopy, such as: basic enzyme kinetics, ligand binding, data handling, and the more recently established interest in the study of protein and DNA stability
Table of Contents
List of protocols
Abbreviations
1. Introduction to light absorption; visible and ultraviolet spectra,
2. Fluorescence principles and measurement,
3. Time resolved fluorescence and phosphorescence spectroscopy,
4. An introduction to circular dichroism,
5. Quantitative determination of equilibrium binding isotherms for the multiple ligand - macromolecule interactions using spectroscopic methods,
6. Steady state kinetics,
7. Spectrophotometric assays,
8. Stopped flow spectroscopy,
9. Stopped flow fluorescence spectroscopy,
10. Stopped flow circular dichroism,
11. Spectrophotometry and fluorimetry of cellular compartments and intracellular processes,
12. Use of optical spectroscopic methods to study the thermodynamic stability of proteins,
13. The use of spectroscopic techniques in the study of DNA stability,
List of suppliers
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?