EMAIL THIS PAGE TO A FRIEND
S1323 Sigma

SID7969543

≥98% (HPLC), solid

Synonym: 2-[2-[(2,3-Dihydrobenzo[1,4]dioxin-6-ylcarbamoyl)methyl]-1-oxo-1,2-dihydroisoquinolin-5-yloxy]propionic acid ethyl ester

  • CAS Number 868224-64-0

  • Empirical Formula (Hill Notation) C24H24N2O7

  • Molecular Weight 452.46

  •  MDL number MFCD04473853

Purchase

Properties

Related Categories Bioactive Small Molecule Alphabetical Index, Bioactive Small Molecules, Cell Biology, Cell Signaling and Neuroscience, Gene Regulation,
InChI Key   KWMBIIQCLUIHDI-UHFFFAOYSA-N
assay   ≥98% (HPLC)
form   solid
color   white to off-white
solubility   DMSO: >10 mg/mL
storage temp.   2-8°C

Description

Biochem/physiol Actions

SID7969543 is a SF-1 (or NR5A1) inhibitor. The Steroidogenic Factor 1 (SF-1, also known as NR5A1) is a transcription factor belonging to the nuclear receptor superfamily. It plays a central role in sex determination and the formation of steroidogenic tissues during development, and is involved in endocrine function throughout life. It has also been reported to have a potential role in obesity. It has also been observed that an increased concentration of SF-1 causes adrenocortical cell proliferation and cancer.

Price and Availability


DISCOVER Bioactive Small Molecules

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?