• USA Home
  • S1390 - Anti-Spectrin antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
S1390 Sigma

Anti-Spectrin antibody produced in rabbit

whole antiserum

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to Actin-associated Proteins/Myosin, Antibodies to Cell and Organelle Proteins,
species reactivity   chicken
application(s)   western blot: 1:400 using total chicken erythrocyte extrac
clone   polyclonal
antibody form   whole antiserum
contains   15 mM sodium azide
shipped in   dry ice
storage temp.   −20°C
Gene Information   chicken ... SPTAN1(374234)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

spectrin from chicken erythrocytes.

Specificity

The antibody specifically stains the α and β chains of total chicken erythrocyte spectrin.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

WGK Germany  -
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?