• USA Home
  • S1435 - Silicone isolators, Press-to-Seal

EMAIL THIS PAGE TO A FRIEND
S1435 Sigma

Silicone isolators, Press-to-Seal

1 well, D × L × W 2.5 mm × 32 mm × 19 mm, silicone/adhesive, oblong

Purchase

Properties

Related Categories Labware, Light Microscopy, Microscopy & Staining More...
material   oblong
  silicone/adhesive
mfr. no.   Grace Bio-Labs
D × L × W   2.5 mm × 32 mm × 19 mm

Description

Application

Isolate specimens on a microscope slide with hydrophobic press-to-seal (removable) or peel-and-stick (adhesive) barriers. Available in silicone thickness 0.5 to 2.5mm as pre-cut gaskets and sheet material; with or without secure seal clean release adhesive on one or both surfaces. Non-adhesive isolators are autoclavable and re-usable for cell culture applications. NOTE: Add approximately 0.15mm to depth per layer of adhesive.

Price and Availability


Tissue Diagnostics
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

GBL665303 Grace Bio-Labs Press-To-Seal silicone isolator, PSA one side, 19 mm x 32 mm x 2.5 mm depth

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?