• USA Home
  • S159 - R(−)-SCH-12679 maleate salt

EMAIL THIS PAGE TO A FRIEND
S159 Sigma

R(−)-SCH-12679 maleate salt

solid

Synonym: R(−)-1-Phenyl-2,3,4,5-tetrahydro-1H-7,8-dimethoxy-3-benzazepine

Purchase

Properties

Related Categories Antagonists, Cell Biology, Cell Signaling and Neuroscience, Dopaminergics, Neuroscience,
InChI Key   TYNKKGLBKXZIHX-XLOMBBFOSA-N
form   solid
optical activity   [α]22/D −11.6°, c = 1.1 in H2O(lit.)
color   white
solubility   DMSO: soluble14 mg/mL
  H2O: soluble17.3 mg/mL
  ethanol: soluble4 mg/mL
  0.1 M HCl: >40 mg/mL
Gene Information   human ... DRD1(1812)

Description

Legal Information

Sold with the permission of Schering-Plough.

Application

R(-)-SCH-12679 maleate salt may be used in D1-dopamine receptor-mediated cell signaling studies.

Biochem/physiol Actions

SCH-12679 is a benzazepine compound and a psychoactive drug that antagonizes D1-dopamine receptor. It reduces the D1-induced locomotor activity and self-mutilation in rats. SCH-12679 decreased aggressiveness and hyperactive behavior in mentally deficient patients.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?