• USA Home
  • S1640 - Anti-Secretory Component (IgA) (Bound and Free) antibody produced in goat

EMAIL THIS PAGE TO A FRIEND
S1640 Sigma

Anti-Secretory Component (IgA) (Bound and Free) antibody produced in goat

whole antiserum

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Immune Cell Signaling and Blood, Antibodies to Human Plasma Proteins,
biological source   goat
antibody form   whole antiserum
clone   polyclonal
contains   15 mM sodium azide
species reactivity   human
application(s)   indirect ELISA: 1:5,000
  quantitative precipitin assay: 1.6-2.4 mg/mL
conjugate   unconjugated
shipped in   dry ice
storage temp.   −20°C

Description

Immunogen

secretory component from human IgA.

Application

Anti-Secretory Component (IgA) (Bound and Free) antibody produced in goat was used in immunoblotting of Pneumococcal protein extracts.

Biochem/physiol Actions

IgA antibody is secretory antibody and is present abundantly in mucous linings of gastrointestinal, respiratory and genitourinary tracts, tears, colostrum and saliva. Thus secretory IgA contributes to the humoral defense mechanism against the pathogens on mucosal surfaces.

Price and Availability


Detect Your Unseen Protein
Safety & Documentation
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer - Over 80,000 Products

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?