• USA Home
  • S1687 - Monoclonal Anti-SLP-76 antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
S1687 Sigma

Monoclonal Anti-SLP-76 antibody produced in mouse

clone AS55, purified immunoglobulin, buffered aqueous solution

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Kinase/Phosphatase Biology, Primary Antibodies,
species reactivity   human
application(s)   immunoprecipitation: 1-5 μg/mL using Jurkat cell lysates
  western blot: 1 μg/mL using Jurkat cell lysates
clone   AS55, monoclonal
antibody form   purified immunoglobulin
form   buffered aqueous solution
isotype   IgG2a
mol wt   antigen mol wt 76 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... LCP2(3937)
biological source   mouse
conjugate   unconjugated

Description

Immunogen

unique peptide corresponding to a portion of human SLP-76 (SH2 [Src Homology 2] domain-leukocyte protein of 76 kDa).

General description

SLP-76 (SH2 domain-containing leukocyte phosphoprotein) is a 76kD protein that is expressed mainly in cells of hematopoietic origin. It facilitates TCR signalling and has a pivotal role in TCR-induced tyrosine phosphorylation of phospholipase Cγ, inositol phosphate accumulation, Ca2+ mobilization, and ras activation. Monoclonal anti-SLP-76 antibody can be used in immunoprecipitation. Mouse anti-SLP-76 antibody reacts specifically with human SLP-76 protein.

Physical form

Solution of ~0.5 mg/ml in phosphate buffered saline containing 0.1% sodium azide.

Preparation Note

Purified from tissue culture supernatant using Protein G.

Application

Monoclonal anti-SLP-76 antibody can be used in western blotting to detect SLP-76 protein using Jurkat cell lysate.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
WGK Germany  3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?