• USA Home
  • S1690 - Multislip cell culture coverslip inserts
EMAIL THIS PAGE TO A FRIEND
S1690 Sigma
Multislip cell culture coverslip inserts
L × W 12 mm × 12 mm
Synonym: cell inserts, coverslips, plate inserts
Purchase
Properties
Related Categories Cell Chamber Slides and Glass Coverslips, Cell Culture Supplies, Labware More...
packaging   pk of 150 coverslips
mfr. no.   Grace Bio-Labs
L × W   12 mm × 12 mm
Description
General description
MultiSlip inserts with 8 (18 mm × 18 mm) or 15 (12 mm × 12 mm) No. 11/2 German Glass cover slips per insert are sterile and ready to use in conventional 86 mm × 128 mm culture plates. Ideally suited for the culture of cells where pre-treatment of the glass surface with a biological coating is recommended.
Simply add sterile solution to the plate, incubate and aspirate. Coating is applied evenly to one side of the glass only, with no overlapping, handling with forceps, or breakage.
Legal Information
Multislip is a trademark of Grace Bio-Labs, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?