• USA Home
  • S1900 - Storage rack for β-emitters, acrylic
EMAIL THIS PAGE TO A FRIEND
S1900 Sigma
Storage rack for β-emitters, acrylic
capacity, 10 1.5-mL microcentrifuge tubes
Purchase
Properties
Related Categories Labware, Radiation Safety, Radiation Safety Racks, Safety Equipment
material   3/8″ acrylic box, lid, and holder
feature   capacity 10 1.5-mL microcentrifuge tubes
availability   not available in EU
Description
General description
Three-part with removable centrifuge tube holder. Box and lid form interlocking seal, minimizing radiation leakage.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?