• USA Home
  • S199 - Sphingosylphosphorylcholine

EMAIL THIS PAGE TO A FRIEND
S199 Sigma

Sphingosylphosphorylcholine

powder

Synonym: (2S,3R,4E)-2-Amino-4-octadecene-3-hydroxy-1-phosphocholine, 2-[[[(2-Amino-3-hydroxy-4-octadecenyl)oxy]hydroxy-phosphinyl]oxy]-N,N,N-trimethylethanaminium inner salt, D-erythro-Sphingosine phosphocholine, Lyso SM (d18:1), Lysosphingomyelin

  • CAS Number 1670-26-4

  • Empirical Formula (Hill Notation) C23H49N2O5P

  • Molecular Weight 464.62

  •  Beilstein Registry Number 7342520

  •  MDL number MFCD02259287

Purchase

Properties

InChI Key   JLVSPVFPBBFMBE-HXSWCURESA-N
form   neat
  powder
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
color   white
solubility   H2O: soluble10 mg/mL
format   neat
storage temp.   −20°C

Description

Biochem/physiol Actions

Sphingosine derivative; increases intracellular Ca2+.

Caution

Hygroscopic

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

S4257 Sphingosylphosphorylcholine, ≥98%, powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?