• USA Home
  • S2009 - Spotting guide and Rf reader
EMAIL THIS PAGE TO A FRIEND
S2009 Sigma
Spotting guide and Rf reader
Spotting guide and Rf reader
Purchase
Description
General description
Place over the spots on TLC plates after development and detection for quick and easy Rf measurements. Markings at the bottom of the template are useful guides for applying samples at uniform distances across the plate. The guide is printed on 5 mm clear acetate.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?