EMAIL THIS PAGE TO A FRIEND
S2145 Sigma

Sera human

Male, Type AB

Purchase

Properties

storage temp.   2-8°C

Description

Preparation Note

Prepared from normal male type AB plasma.

Application

Human AB serum is used in tissue engineering, transplantation and cell therapy applications for the expansion of mesenchymal stem cells (MSC) from adipose tissue or mesenchymal stromal cells from human bone marrow; for standardized limbal epithelial stem cell graft generation and transplantation; for ex vivo expansion of NK cells from peripheral blood in Hematopoeitic Stem Cell Expansion Medium; and for upgrading pretransplant human islet culture technology.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

recommended

Product #

Description

Add to Cart

S0189 Stemline® Hematopoietic Stem Cell Expansion Medium, Serum-free, Without L-glutamine, liquid, sterile-filtered, suitable for cell culture
S0192 Stemline® II Hematopoietic Stem Cell Expansion Medium, Serum-free, contains L-glutamine, liquid, sterile-filtered, suitable for cell culture
S1569 Stemline® Mesenchymal Stem Cell Expansion Medium, Without L-glutamine, liquid, sterile-filtered, suitable for cell culture

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?