EMAIL THIS PAGE TO A FRIEND
S2278 Sigma
Stirred cells
Purchase
Description
Features and Benefits
Autoclavable, low pressure cells are designed for ultra- or microfiltration with pressure not exceeding 70 psi. Features include a vent/relief valve and a magnetic stirring bar to minimize concentration polarization of the membrane surface. All units come complete with 4 feet of polyethylene tubing, a hose barb for filtrate and a 1/4 in. nickel-plated brass fitting for connection to an external regulated pressure source.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?