• USA Home
  • S2314 - δ-Aminovaleryl-[Pro9, Leu(N-Me)10]-Substance P Fragment 7-11

EMAIL THIS PAGE TO A FRIEND
S2314 Sigma

δ-Aminovaleryl-[Pro9, Leu(N-Me)10]-Substance P Fragment 7-11

>99% (HPLC), solid

  • Empirical Formula (Hill Notation) C40H59N7O6S

  • Molecular Weight 766.00

  •  MDL number MFCD00144411

  •  PubChem Substance ID 329824518

Purchase

Properties

Related Categories Agonist, Cell Biology, Cell Signaling and Neuroscience, Neurotransmission (Obesity), Obesity Research,
InChI Key   HQKPTSSZOJLFBZ-LJADHVKFSA-N
assay   >99% (HPLC)
form   solid
storage temp.   −20°C

Description

Amino Acid Sequence

δ-Ava-Phe-Phe-Pro-N-Me-Leu-Met-NH2

Biochem/physiol Actions

Selective receptor agonist for NK1.

Price and Availability


Amplified Detection

All labs need water
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?