• USA Home
  • S2437 - SCF Soluble Receptor human

EMAIL THIS PAGE TO A FRIEND
S2437 Sigma

SCF Soluble Receptor human

>95% (SDS-PAGE), recombinant, expressed in baculovirus infected Sf21 cells, lyophilized powder

Synonym: CD117, SCFsR, Stem Cell Factor Soluble Receptor

Purchase

Description

Analysis Note

The bioactivity is measured by its ability to inhibit SCF-dependent proliferation of TF-1 cells.

Physical form

Lyophilized from a 0.2 μm filtered solution in PBS, pH 7.4, with 5% trehalose and containing 50 μg of BSA per 1 μg cytokine

Biochem/physiol Actions

Recombinant human stem cell factor soluble receptor (CD117, the gene product of the c-kit proto-oncogene) is a member of the class III subfamily of receptor tyrosine kinases. It binds SCF with high affinity and induces receptor homodimerization and signal transduction.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?