• USA Home
  • S2639 - Sigma Library of FT-IR Spectra, Volumes 1 and 2

EMAIL THIS PAGE TO A FRIEND
S2639 Sigma

Sigma Library of FT-IR Spectra, Volumes 1 and 2

set of 1

  •  ISBN-10 0-941633-10-1

  •  ISBN-13 978-0-941633-10-9

Purchase

Properties

Related Categories Books and Software, Labware, Spectroscopy Books More...
publication info   R.J. Keller, Sigma Chemical Company, MO, 1986, 2,894 pp., hard cover
packaging   set of 1

Description

General description

This set contains over 10,400 FT-IR spectra of biochemicals and related organics. Spectra are linear in wavenumber with 2 cm -1 resolution, measure 18 × 4.5 cm and are arranged by categories.

Table of Contents

VOLUME I
Amino Acids, Analogs and Derivatives
Peptides
Enzymes
Substrates
Proteins
Toxins
Alkaloids
Chemotherapeutic Agents
Controlled Substances
Nucleosides
Nucleotides
Coenzymes
Oligo- and Polynucleotides
Purines and Pyrimidines
Carbohydrates, Analogs and Derivatives
Polysaccharides
Steroids
Alcohols
Fatty Acids, Esters, and Acid Chlorides
Oils, Gums, and Waxes
Glycerides
Phosphos- and Sphingolipids
VOLUME II
Biogenic Amines
Porphyrins
Biological Pigments
Plant Growth Regulators
Buffers
Surfactants
Dyes and Stains
Organics
Silanes
Deuterated Chemicals
Inorganics
Acetoen Powders and Miscellaneous
Alphabetical Index
Molecular Formula Index
CAS Number Index

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?