• USA Home
  • S2645 - Spirochetes TISSUE-TROL

EMAIL THIS PAGE TO A FRIEND
S2645 Sigma

Spirochetes TISSUE-TROL

from rabbit testicle tissue containing spirochetes (Treponema pallidum)

Purchase

Properties

Related Categories Cell Biology, Hematology and Histology, TISSUE-TROL Control Slides More...
Quality Level   GMP-IVD
biological source   from rabbit testicle tissue containing spirochetes (Treponema pallidum)
shelf life   Expiry date on the label.
IVD   for in vitro diagnostic use

Description

Components

25 slides per box. 1 stained slide is provided with each box. The stained slide has been stained with Sigma Procedure No. HT101

Application

Suitable for use with Sigma Silver Stain, Modified Steiner-Steiner, Procedure No. HT101 and other silver impregnation techniques.

Legal Information

TISSUE-TROL is a trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

TTR011 Spirochetes TISSUE-TROL Control Slides, from rabbit testicle tissue containing spirochetes (Treponema pallidum)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?