EMAIL THIS PAGE TO A FRIEND
S2875 Sigma

Dibutyl sebacate

≥90%

Synonym: Decanedioic acid dibutyl ester, Sebacic acid dibutyl ester

  • CAS Number 109-43-3

  • Linear Formula [-(CH2)4CO2(CH2)3CH3]2

  • Molecular Weight 314.46

  •  Beilstein Registry Number 1798308

  •  EC Number 203-672-5

  •  MDL number MFCD00027218

  •  PubChem Substance ID 24899536

Purchase

Properties

Related Categories Biochemicals and Reagents, Lipids, Other Lipid Related Products, Other Lipids & Mixtures of Lipids
InChI Key   PYGXAGIECVVIOZ-UHFFFAOYSA-N
assay   ≥90%
form   liquid
refractive index   n20/D 1.441(lit.)
bp   178-179 °C/3 mmHg(lit.)
density   0.936 g/mL at 25 °C(lit.)

Price and Availability


The Future of Bioimaging
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
VS1150000
Flash Point(F) 
352.4 °F
Flash Point(C) 
178 °C

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?