EMAIL THIS PAGE TO A FRIEND
S3012 Sigma

Sera from coyote

lyophilized powder

Purchase

Properties

Related Categories Biochemicals and Reagents, Plasma & Blood Proteins, Plasma, Blood, and Related Proteins and Reagents, Proteins and Derivatives, Sera More...
form   lyophilized powder
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?