• USA Home
  • S3190 - SigmaScreen Collagen I coated plates
EMAIL THIS PAGE TO A FRIEND
S3190 Sigma
SigmaScreen Collagen I coated plates
96-well, black/clear bottom
Purchase
Properties
Related Categories Cell Culture, Coated Plates, Drug Discovery More...
material   polystyrene
description   (flat bottom)
storage temp.   2-8°C
Description
General description
SigmaScreen Collagen I coated multiwell plates are coated with Collagen I from rat tail. Collagen I, a fibrous extracellular matrix protein, is found in tendons, bones, and a variety of tissues and organs. Commonly used as an attachment factor for serum free or low serum cultures, cells attach firmly to Collagen I via integrin surface receptors, thereby promoting cell attachment and spreading. SigmaScreen Collagen I coated multiwell plates can be used with various cell lines including COS-7 and PC-12. For a more comprehensive listing of cell lines used with Collagen I coated multiwell plates, please refer to the Collagen I coated plate technical bulletin. Manufactured under ISO9002 in Sigma′s GMP facility, Sigma′s Collagen I coated multiwell plates are bioburden tested (fungal and bacterial) and endotoxin free.
Legal Information
SigmaScreen is a trademark of Sigma-Aldrich Co. LLC
Price and Availability
All labs need water
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?