• USA Home
  • S3312 - Panorama® Array Stripping Solution

EMAIL THIS PAGE TO A FRIEND
S3312 Sigma

Panorama® Array Stripping Solution

1 ×, ready-to-use solution

Purchase

Properties

form   ready-to-use solution
concentration   1 ×
foreign activity   DNase, RNase, none detected

Description

Application

Convenient, ready-to-use stripping solution optimized for stripping Panorama Gene Arrays. Each lot is made with Molecular Biology grade reagents and is performance and quality control tested. Enough solution to strip 4 arrays when following the Panorama Gene Array protocol.

Legal Information

Panorama is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?