• USA Home
  • S3527 - Workshields, acrylic, for β-emitters
EMAIL THIS PAGE TO A FRIEND
S3527 Sigma
Workshields, acrylic, for β-emitters
size 16 in. × 16 in.
Purchase
Properties
Related Categories Labware, Radiation Safety, Radiation Safety Workshields, Safety Equipment
size   16 in. × 16 in.
thickness   3/8 in. , with removable pedestal style feet
Description
General description
Free-standing shields of clear acrylic provide effective protection against most β-emitting isotopes, including 32P. Angled fronts permit easy viewing.
Intended to be positioned at the side or behind the work area to protect other laboratory workers from exposure. Sets up vertically or at a 75° angle so reflections can be eliminated.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?