• USA Home
  • S3641 - [D-Arg1, D-Trp7,9, Leu11]-Substance P

EMAIL THIS PAGE TO A FRIEND
S3641 Sigma

[D-Arg1, D-Trp7,9, Leu11]-Substance P

≥97% (HPLC)

Synonym: Spantide I

  • CAS Number 91224-37-2

  • Empirical Formula (Hill Notation) C75H108N20O13

  • Molecular Weight 1497.79

  •  MDL number MFCD00076805

Purchase

Properties

Related Categories Antagonist, Cell Biology, Cell Signaling and Neuroscience, Peptides and Proteins, Peptides for Cell Biology,
InChI Key   SAPCBHPQLCZCDV-CHPWDEGWSA-N
assay   ≥97% (HPLC)
storage temp.   −20°C
Gene Information   human ... TAC1(6863), TACR1(6869)
mouse ... TAC1(21333), TACR1(21336)
rat ... TAC1(24806), TACR1(24807)

Description

Amino Acid Sequence

D-Arg-Pro-Lys-Pro-Gln-Gln-D-Trp-Phe-D-Trp-Leu-Leu-NH2

Biochem/physiol Actions

Potent substance P antagonist with weak histamine-releasing action

Price and Availability


Detect Your Unseen Protein

All labs need water

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?