• USA Home
  • S3651 - Storage container, acrylic, for β-emitters
EMAIL THIS PAGE TO A FRIEND
S3651 Sigma
Storage container, acrylic, for β-emitters
L × W × H 4 1/2 in. × 3 1/2 in. × 2 in.
Purchase
Properties
Related Categories Labware, Radiation Safety, Radiation Safety Containers, Safety Equipment
L × W × H   4 1/2 in. × 3 1/2 in. × 2 in.
Description
General description
Made from 1/2 in. clear acrylic with fitted lid for shielded storage of most β-emitters, including 32P.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?