• USA Home
  • S3657 - Reciprocating / orbital shaker, AROS
EMAIL THIS PAGE TO A FRIEND
S3657 Sigma
Reciprocating / orbital shaker, AROS
AC/DC input 240 V AC, 0.18 A
Purchase
Description
General description
Fully adjustable from very slow agitation to vigorous vortexing action. Digital speed control is settable from 12 to 280 rpm. Mixing action is changed with a simple adjustment tool (included) for linear or orbital movements of 0.63, 0.75, 1.0, or 1.25 in.
Mixing surface is 11.7 × 14 in. (30 × 35 cm) silicone rubber pad with raised edges for secure use with staining tray or similar vessel. Very low power consumption allows use in incubator without over-heating.
Legal Information
AROS is a trademark of Barnstead/Thermolyne Corp.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?