• USA Home
  • S3764 - Sorbitol Dehydrogenase from sheep liver

EMAIL THIS PAGE TO A FRIEND
S3764 Sigma

Sorbitol Dehydrogenase from sheep liver

lyophilized powder, ≥20 units/mg protein

Synonym: L-Iditol Dehydrogenase, L-Iditol:NAD+ 5-oxidoreductase, Polyol Dehydrogenase, SDH

Purchase

Properties

Related Categories 1.1.x.x Acting on hydroxyl groups, 1.x.x.x Oxidoreductases, Biochemicals and Reagents, Carbohydrate Metabolism, Cell Biology,
form   lyophilized powder
composition   Protein, 5-25% biuret
foreign activity   GLDH, LDH, and MDH ≤0.05%
storage temp.   −20°C

Description

General description

Activity of sorbitol dehydrogenase increases in diabetic animals.

Unit Definition

One unit will convert 1.0 μmole of D-fructose to D-sorbitol per min at pH 7.6 at 25 °C.

Physical form

Contains maltose.

Application

Sorbitol dehydrogenase has been used in a study to investigate osmotic stress induced oxidative damage as a possible mechanism of cataract formation in diabetes.

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

92210 Timestrip Plus 0 °C
06693 Timestrip Plus -20 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?