• USA Home
  • S3776 - Storage rack for β-emitters, acrylic
EMAIL THIS PAGE TO A FRIEND
S3776 Sigma
Storage rack for β-emitters, acrylic
capacity, 24 1.5-mL tubes, with removable block-style insert and fitted lid
Purchase
Properties
Related Categories Labware, Radiation Safety, Radiation Safety Racks, Safety Equipment
feature   capacity 24 1.5-mL tubes
Description
General description
When assembled, at least 1/2 in. of acrylic protects user from radiation.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?