• USA Home
  • S3806 - Soybean casein digest agar

EMAIL THIS PAGE TO A FRIEND
S3806 Sigma

Soybean casein digest agar

Synonym: General purpose medium

Purchase

Properties

pH   7.3±0.2 (25 °C)

Description

Application

General purpose medium used for the cultivation of a wide variety of microorganisms.

Other Notes

Components (g/L)
Casein Enzymic Hydrolysate 15.0
Papaic Digest of Soyabean Meal 5.0
Sodium Chloride 5.0
Agar 15.0

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

22091 Tryptic Soy Agar, for microbiology
22095 CASO Agar, for microbiology

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?