• USA Home
  • S3939 - Staurosporine aglycone

EMAIL THIS PAGE TO A FRIEND
S3939 Sigma

Staurosporine aglycone

≥95%

Synonym: K-252c

Purchase

Description

Biochem/physiol Actions

K-252c is a PKC inhibitor active at submicromolar concentrations.

Staurosporine is a microbial alkaloid that inhibits protein kinase C (PKC) but lacks selectivity and may inhibit other protein kinases. Substitution of one of the indole nitrogen atoms by non-glycosidic indolocarbazoles lacking aminoalkyl side chain results in Staurosporine aglycone with increased selectivity for PKC. Staurosporine aglycone inhibits the autophosphorylation of c-Met and activation of MAPK and Akt in cancer cells.

Packaging

1, 5 mg in glass bottle

Application

RGC-5 cells were treated with Staurosporine aglycone to study the effect of staurosporine analogs on the process of neuronal differentiation.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

related product

Product #

Description

Add to Cart

S3939 Staurosporine aglycone, ≥95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?