EMAIL THIS PAGE TO A FRIEND
S4016 Sigma

Solvent Yellow 7

Synonym: p-Phenylazophenol

  •  Colour Index Number 11800

  •  EC Number 216-880-6

Purchase

Properties

composition   Dye content, ~98%

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
SM8300000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?