• USA Home
  • S4146 - Sarafotoxin S6b Atractaspis engaddensis sequence

EMAIL THIS PAGE TO A FRIEND
S4146 Sigma

Sarafotoxin S6b Atractaspis engaddensis sequence

≥90% (HPLC)

Synonym: SRTX-b

  • CAS Number 116303-65-2

  • Empirical Formula (Hill Notation) C110H159N27O34S5

  • Molecular Weight 2563.92

  •  MDL number MFCD00153553

Purchase

Description

Amino Acid Sequence

Cys-Ser-Cys-Lys-Asp-Met-Thr-Asp-Lys-Glu-Cys-Leu-Tyr-Phe-Cys-His-Gln-Asp-Val-Ile-Trp [Disulfide Bridges: 1-15, 3-11]

Biochem/physiol Actions

ETA agonist; increases intracellular Ca2+

Price and Availability


Making Headway

All labs need water

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?