• USA Home
  • S4384 - Sarcosine Dehydrogenase from Pseudomonas sp.

EMAIL THIS PAGE TO A FRIEND
S4384 Sigma

Sarcosine Dehydrogenase from Pseudomonas sp.

lyophilized powder, 0.5-1.5 units/mg protein

Synonym: Sarcosine:(acceptor) oxidoreductase (demethylating)

Purchase

Description

Unit Definition

One unit will convert 1.0 μmole of sarcosine to glycine and formaldehyde per min at pH 7.5 at 37 °C.

Physical form

Lyophilized powder containing approx. 60% sucrose, 10% potassium phosphate buffer salts and trace EDTA

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?